Merck
The atypical homeodomain transcription factor Mohawk controls tendon morphogenesis.
Molecular and cellular biology (2010-08-11)
Wenjin Liu, Spencer S Watson, Yu Lan, Douglas R Keene, Catherine E Ovitt, Han Liu, Ronen Schweitzer, Rulang Jiang
ZUSAMMENFASSUNG
The Mohawk homeobox (Mkx) gene encodes a new atypical homeodomain-containing protein with transcriptional repressor activity. Mkx mRNA exhibited dynamic expression patterns during development of the palate, somite, kidney, and testis, suggesting that it may be an important regulator of multiple developmental processes. To investigate the roles of Mkx in organogenesis, we generated mice carrying a null mutation in this gene. Mkx(-/-) mice survive postnatally and exhibit a unique wavy-tail phenotype. Close examination revealed that the mutant mice had smaller tendons than wild-type littermates and that the rapid postnatal growth of collagen fibrils in tendons was disrupted in Mkx(-/-) mice. Defects in tendon development were detected in the mutant mouse embryos as early as embryonic day 16.5 (E16.5). Although collagen fibril assembly initially appeared normal, the tendons of Mkx(-/-) embryos expressed significantly reduced amounts of collagen I, fibromodulin, and tenomodulin in comparison with control littermates. We found that Mkx mRNA was strongly expressed in differentiating tendon cells during embryogenesis and in the tendon sheath cells in postnatal stages. In addition to defects in tendon collagen fibrillogenesis, Mkx(-/-) mutant mice exhibited abnormal tendon sheaths. These results identify Mkx as an important regulator of tendon development.