Merck
  • Developmentally dynamic colocalization patterns of DSCAM with adhesion and synaptic proteins in the mouse retina.
Developmentally dynamic colocalization patterns of DSCAM with adhesion and synaptic proteins in the mouse retina.
Molecular vision (2014-10-30)
Gabriel Belem de Andrade, Landon Kunzelman, Morgan M Merrill, Peter G Fuerst
ZUSAMMENFASSUNG
The Down syndrome cell adhesion molecule (Dscam) gene is required for normal dendrite arborization and lamination in the mouse retina. In this study, we characterized the developmental localization of the DSCAM protein to better understand the postnatal stages of retinal development during which laminar disorganization occur in the absence of the protein. Immunohistochemistry and colocalization analysis software were used to assay the localization of the DSCAM protein during development of the retina. We found that DSCAM was initially localized diffusely throughout mouse retinal neurites but then adopted a punctate distribution. DSCAM colocalized with catenins in the adult retina but was not detected at the active zone of chemical synapses, electrical synapses, and tight junctions. Further analysis identified a wave of colocalization between DSCAM and numerous synaptic and junction proteins coinciding with synaptogenesis between bipolar and retinal ganglion cells. Research presented in this study expands our understanding of DSCAM function by characterizing its location during the development of the retina and identifies temporally regulated localization patterns as an important consideration in understanding the function of adhesion molecules in neural development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Connexin 35/36 Antikörper, Klon 8F6.2, clone 8F6.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Growth Associated Protein 43-Antikörper, Klon 9-1E12, clone 9-1E12, Chemicon®, from mouse
Sigma-Aldrich
Anti-NMDAR2 A/B-Antikörper, pan, Chemicon®, from rabbit
Sigma-Aldrich
Anti-MAGI2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Syntaxin-4-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
Anti-MAGI3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution