Merck
  • Diagnostic cytology seminar. 23rd annual scientific meeting American Society of Cytology San Juan, Puerto Rico November 8, 1975.
Diagnostic cytology seminar. 23rd annual scientific meeting American Society of Cytology San Juan, Puerto Rico November 8, 1975.
Acta cytologica (1976-09-01)
PMID185868