Merck
  • Detailed features in the local mode overtone bands of ethane, neopentane, tetramethylbutane, and hexamethylbenzene.
Detailed features in the local mode overtone bands of ethane, neopentane, tetramethylbutane, and hexamethylbenzene.
J. Chem. Phys. (1980)
Henry BR and Greenlay WRA.