Merck
  • The human RIL gene: mapping to human chromosome 5q31.1, genomic organization and alternative transcripts.
The human RIL gene: mapping to human chromosome 5q31.1, genomic organization and alternative transcripts.
Gene (1998-05-09)
A A Bashirova, M L Markelov, T V Shlykova, E V Levshenkova, R A Alibaeva, E I Frolova
ZUSAMMENFASSUNG
The ril gene encoding a LIM domain protein of an unknown function was previously identified by differential expression cloning as a candidate tumor suppressor gene in rat fibroblasts (Kiess, M., Scharm, B., Aguzzi, A., Hajnal, A., Klemenz, R., Schwarte-Waldhoff, I., Schafer, R., 1995. Expression of ril, a novel LIM domain gene, is down-regulated in HRAS-transformed cells and restored in phenotypic revertants. Oncogene 10, 61-68). Searching for novel genes on human chromosome 5q31.1 by the cDNA selection technique, we isolated a cDNA clone identical with the cDNA of the human RIL gene (GenBank Accession No. X93510). The human 5q31.1 region is of interest because it contains the cytokine gene cluster and is frequently deleted in the malignant cells of patients with myelodysplasia and myeloid leukemia. Using Southern blot analysis and restriction mapping of genomic YAC (yeast artificial chromosome) and cosmid clones, we located the human RIL gene 240-260 kb telomeric to the IRF1 gene and characterized its genomic structure. PCR analysis indicated the presence of two alternative RIL transcripts in human fetal brain mRNA. The major transcript is identical with the RIL cDNA previously deposited in GenBank and contains seven exons distributed over 14.5 kb of genomic DNA with the two last 3'-exons coding a LIM domain. The minor transcript lacks the sixth exon compared with the major transcript, which leads to the loss of the LIM domain. We also identified two putative transcription start points (tsp) and sequenced the 5'-flanking region of RIL to reveal potential binding sites for transcriptional factors.