Merck
  • Solid state deoximation with ammonium persulfate-silica gel: Regeneration of carbonyl compounds using microwaves.
Solid state deoximation with ammonium persulfate-silica gel: Regeneration of carbonyl compounds using microwaves.
Tetrahedron Letters (1997)
Varma RS and Meshram HM.