Merck
  • Ligand-optimized electroless synthesis of silver nanotubes and their activity in the reduction of 4-nitrophenol.
Ligand-optimized electroless synthesis of silver nanotubes and their activity in the reduction of 4-nitrophenol.
Nanotechnology (2011)
Muench F, et al.