Merck
Traces of phosgene in chloroform: consequences for extraction of anthracyclines.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2006-11-23)
Kristof E Maudens, Sarah M R Wille, Willy E Lambert
ZUSAMMENFASSUNG
Chloroform is commonly used to extract anthracyclines from various biological matrices. However, their determination can be seriously compromised by phosgene traces present as a result of failing stabilization of chloroform. Out of the three varieties in which chloroform exists (not stabilized, stabilized with an alcohol and stabilized with a hydrocarbon) only the ethanol stabilized type minimizes chances on creating artifacts. Chromatographic separation after extraction of four anthracyclines (doxorubicin, epirubicin, daunorubicin and idarubicin) and two metabolites (13-S-dihydrodoxorubicin and 13-S-dihydroepirubicin) with chloroform under various conditions indicate that the appropriate choice of stabilizer in this extraction solvent is highly relevant.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer