Merck
  • Simple liquid-liquid partition system for isolation of labeled oleic acid from mixtures with glycerides.
Simple liquid-liquid partition system for isolation of labeled oleic acid from mixtures with glycerides.
Journal of lipid research (1969-05-01)
P Belfrage, M Vaughan
ZUSAMMENFASSUNG
Oleic acid has a partition coefficient, upper phase/lower phase, of 1.9 (22 degrees C) in the liquid-liquid partition system described herein. Tri-, di-, and monoolein are found almost exclusively in the lower (organic) phase. Oleic acid can be quantitatively removed from mixtures of triglyceride and partial glycerides by means of this partition system under conditions resembling those in a lipase assay.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)