Merck
  • [Technical recommendations and best practice guidelines for May-Grünwald-Giemsa staining: literature review and insights from the quality assurance].
[Technical recommendations and best practice guidelines for May-Grünwald-Giemsa staining: literature review and insights from the quality assurance].
Annales de pathologie (2015-07-21)
Eric Piaton, Monique Fabre, Isabelle Goubin-Versini, Marie-Françoise Bretz-Grenier, Monique Courtade-Saïdi, Serge Vincent, Geneviève Belleannée, Françoise Thivolet, Jean Boutonnat, Hervé Debaque, Jocelyne Fleury-Feith, Philippe Vielh, Béatrix Cochand-Priollet, Caroline Egelé, Jean-Pierre Bellocq, Jean-François Michiels
ZUSAMMENFASSUNG
May-Grünwald-Giemsa (MGG) stain is a Romanowsky-type, polychromatic stain as those of Giemsa, Leishman and Wright. Apart being the reference method of haematology, it has become a routine stain of diagnostic cytopathology for the study of air-dried preparations (lymph node imprints, centrifuged body fluids and fine needle aspirations). In the context of their actions of promoting the principles of quality assurance in cytopathology, the French Association for Quality Assurance in Anatomic and Cytologic Pathology (AFAQAP) and the French Society of Clinical Cytology (SFCC) conducted a proficiency test on MGG stain in 2013. Results from the test, together with the review of literature data allow pre-analytical and analytical steps of MGG stain to be updated. Recommendations include rapid air-drying of cell preparations/imprints, fixation using either methanol or May-Grünwald alone for 3-10minutes, two-step staining: 50% May-Grünwald in buffer pH 6.8 v/v for 3-5minutes, followed by 10% buffered Giemsa solution for 10-30minutes, and running water for 1-3minutes. Quality evaluation must be performed on red blood cells (RBCs) and leukocytes, not on tumour cells. Under correct pH conditions, RBCs must appear pink-orange (acidophilic) or buff-coloured, neither green nor blue. Leukocyte cytoplasm must be almost transparent, with clearly delineated granules. However, staining may vary somewhat and testing is recommended for automated methods (slide stainers) which remain the standard for reproducibility. Though MGG stain remains the reference stain, Diff-Quik(®) stain can be used for the rapid evaluation of cell samples.