Merck
  • Systems genetics of the lateral septal nucleus in mouse: heritability, genetic control, and covariation with behavioral and morphological traits.
Systems genetics of the lateral septal nucleus in mouse: heritability, genetic control, and covariation with behavioral and morphological traits.
PloS one (2012-09-07)
Alexander Talishinsky, Glenn D Rosen
ZUSAMMENFASSUNG
The lateral septum has strong efferent projections to hypothalamic and midbrain regions, and has been associated with modulation of social behavior, anxiety, fear conditioning, memory-related behaviors, and the mesolimbic reward pathways. Understanding natural variation of lateral septal anatomy and function, as well as its genetic modulation, may provide important insights into individual differences in these evolutionarily important functions. Here we address these issues by using efficient and unbiased stereological probes to estimate the volume of the lateral septum in the BXD line of recombinant inbred mice. Lateral septum volume is a highly variable trait, with a 2.5-fold difference among animals. We find that this trait covaries with a number of behavioral and physiological phenotypes, many of which have already been associated with behaviors modulated by the lateral septum, such as spatial learning, anxiety, and reward-seeking. Heritability of lateral septal volume is moderate (h(2) = 0.52), and much of the heritable variation is caused by a locus on the distal portion of chromosome (Chr) 1. Composite interval analysis identified a secondary interval on Chr 2 that works additively with the Chr 1 locus to increase lateral septum volume. Using bioinformatic resources, we identified plausible candidate genes in both intervals that may influence the volume of this key nucleus, as well as associated behaviors.