Merck
Synthesis and characterization of hybrid MCM-41 materials for heavy metal adsorption.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association (2009)
Dimos K, et al.