Merck
  • Synthesis and modification of low-molecular-weight polybutadiene and polystyrene for use as adhesive additive to asphalt.
Synthesis and modification of low-molecular-weight polybutadiene and polystyrene for use as adhesive additive to asphalt.
Russian Journal of Applied Chemistry (2012)
Pankov IV