Merck
  • Selective inhibition of mutant isocitrate dehydrogenase 1 (IDH1) via disruption of a metal binding network by an allosteric small molecule.
Selective inhibition of mutant isocitrate dehydrogenase 1 (IDH1) via disruption of a metal binding network by an allosteric small molecule.
The Journal of biological chemistry (2014-11-14)
Gejing Deng, Junqing Shen, Ming Yin, Jessica McManus, Magali Mathieu, Patricia Gee, Timothy He, Chaomei Shi, Olivier Bedel, Larry R McLean, Frank Le-Strat, Ying Zhang, Jean-Pierre Marquette, Qiang Gao, Bailin Zhang, Alexey Rak, Dietmar Hoffmann, Eamonn Rooney, Aurelie Vassort, Walter Englaro, Yi Li, Vinod Patel, Francisco Adrian, Stefan Gross, Dmitri Wiederschain, Hong Cheng, Stuart Licht
ZUSAMMENFASSUNG
Cancer-associated point mutations in isocitrate dehydrogenase 1 and 2 (IDH1 and IDH2) confer a neomorphic enzymatic activity: the reduction of α-ketoglutarate to d-2-hydroxyglutaric acid, which is proposed to act as an oncogenic metabolite by inducing hypermethylation of histones and DNA. Although selective inhibitors of mutant IDH1 and IDH2 have been identified and are currently under investigation as potential cancer therapeutics, the mechanistic basis for their selectivity is not yet well understood. A high throughput screen for selective inhibitors of IDH1 bearing the oncogenic mutation R132H identified compound 1, a bis-imidazole phenol that inhibits d-2-hydroxyglutaric acid production in cells. We investigated the mode of inhibition of compound 1 and a previously published IDH1 mutant inhibitor with a different chemical scaffold. Steady-state kinetics and biophysical studies show that both of these compounds selectively inhibit mutant IDH1 by binding to an allosteric site and that inhibition is competitive with respect to Mg(2+). A crystal structure of compound 1 complexed with R132H IDH1 indicates that the inhibitor binds at the dimer interface and makes direct contact with a residue involved in binding of the catalytically essential divalent cation. These results show that targeting a divalent cation binding residue can enable selective inhibition of mutant IDH1 and suggest that differences in magnesium binding between wild-type and mutant enzymes may contribute to the inhibitors' selectivity for the mutant enzyme.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-α-Hydroxyglutarsäure Dinatriumsalz, ≥98.0% (GC)