Merck
  • Sol-gel ormosils doped with TEMPO as recyclable catalysts for the selective oxidation of alcohols.
Sol-gel ormosils doped with TEMPO as recyclable catalysts for the selective oxidation of alcohols.
Advanced Synthesis & Catalysis (2002)
Ciriminna R, et al.