Merck
  • Facile one pot synthesis of 8-chloro-[1, 2, 4] triazolo [4, 3-a] pyrazines via oxidative cyclisation using chloramine T.
Facile one pot synthesis of 8-chloro-[1, 2, 4] triazolo [4, 3-<I>a</I>] pyrazines via oxidative cyclisation using chloramine T.
Tetrahedron Letters (2015)
Mal S, et al.