Merck
  • The quantitative assay of minerals for Fe2+ and Fe3+ using 1, 10-phenanthroline: II. A photochemical method.
The quantitative assay of minerals for Fe<SUP>2+</SUP> and Fe<SUP>3+</SUP> using 1, 10-phenanthroline: II. A photochemical method.
Soil Science Society of America Journal. Soil Science Society of America (1981)
Stucki JW.