Merck
  • C1q/TNF-related protein-3 represents a novel and endogenous lipopolysaccharide antagonist of the adipose tissue.
C1q/TNF-related protein-3 represents a novel and endogenous lipopolysaccharide antagonist of the adipose tissue.
Endocrinology (2010-08-27)
Andrea Kopp, Margarita Bala, Christa Buechler, Werner Falk, Philipp Gross, Markus Neumeier, Jürgen Schölmerich, Andreas Schäffler
ZUSAMMENFASSUNG
Proteins secreted by adipocytes (adipokines) play an important role in the pathophysiology of type 2 diabetes mellitus and the associated chronic and low-grade state of inflammation. It was the aim to characterize the antiinflammatory potential of the new adipocytokine, C1q/TNF-related protein-3 (CTRP-3), which shows structural homologies to the pleiotropic adipocytokine adiponectin. mRNA and protein expression of CTRP-3 was analyzed by RT-PCR and Western blot. Recombinant CTRP-3 and small interfering RNA-based strategies were used to investigate the effect of CTRP-3 on toll-like receptor (TLR) ligand, lipopolysaccharide (LPS)-, and lauric acid-induced chemokine release of monocytes and adipocytes. Together with complex ELISA-based techniques, a designed TLR4/myeloid differentiation protein-2 fusion molecule shown to bind LPS was used to prove the ability of CTRP-3 to act as endogenous LPS antagonist. CTRP-3 is synthesized in monocytes and adipocytes. The recombinant protein dose-dependently inhibits the release of chemokines in monocytes and adipocytes that were induced by lauric acid, LPS, and other TLR ligands in vitro and ex vivo. CTRP-3 inhibits monocyte chemoattractant protein-1 release in adipocytes, whereas small interfering RNA-mediated knockdown of CTRP-3 up-regulates monocyte chemoattractant protein-1 release, reduces lipid droplet size, and decreases intracellular triglyceride concentration in adipocytes, causing a dedifferentiation into a more proinflammatory and immature phenotype. By using a designed TLR4/MD-2 fusion molecule, it is shown by different techniques that CTRP-3 specifically and effectively inhibits the binding of LPS to its receptor, TLR4/MD-2. CTRP-3 inhibits three basic and common proinflammatory pathways involved in obesity and type 2 diabetes mellitus (adipo-inflammation) by acting as an endogenous LPS antagonist of the adipose tissue.