Merck
  • Modeling of photocatalytic mineralization of phthalic acid in TiO2 suspension using response surface methodology (RSM).
Modeling of photocatalytic mineralization of phthalic acid in TiO<SUB>2</SUB> suspension using response surface methodology (RSM).
Desalination and Water Treatment (2015)
Abaamrane A, et al.