Merck
  • A new synthetic procedure for ordered mesoporous γ-alumina using phthalic acid as an interfacial protector.
A new synthetic procedure for ordered mesoporous γ-alumina using phthalic acid as an interfacial protector.
Materials Letters (2011)
Huang F, et al.