Merck
  • Desulfurization of N-Substituted 2-Thioxothiazolidin-4-ones in Aqueous Solutions of Chloroacetic Acid.
Desulfurization of <I>N</I>-Substituted 2-Thioxothiazolidin-4-ones in Aqueous Solutions of Chloroacetic Acid.
Russ. J. Org. Chem. (1998)
Orlinskii MM.