Merck
  • Development of eco-efficient micro-porous membranes via electrospinning and annealing of poly (lactic acid).
Development of eco-efficient micro-porous membranes via electrospinning and annealing of poly (lactic acid).
Journal of Membrane Science (2013)
Li L, et al.