Merck
  • Antioxidant and antimicrobial activities, and phenolic compounds of selected Inula species from Turkey.
Antioxidant and antimicrobial activities, and phenolic compounds of selected Inula species from Turkey.
Natural product communications (2013-06-07)
Alper Gökbulut, Onural Ozhan, Basri Satilmiş, Kadir Batçioğlu, Selami Günal, Engin Sarer
ZUSAMMENFASSUNG
Three Inula species, I. viscosa, I. helenium ssp. turcoracemosa and I. montbretiana, collected from different locations of Anatolia were investigated for their antioxidant and antimicrobial potential, and their total phenolic content and phenolic composition. Antioxidant activities of various extracts of the plant parts were measured using DPPH radical scavenging and ABTS assays. Antimicrobial potential of methanol extracts of the plant parts was determined by the agar dilution method against Staphylococcus aureus, Enterococcus faecalis, Escherichia coli, Pseudomonas aeruginosa, Candida albicans and Candida tropicalis. All the extracts were more active against Gram-positive bacteria and yeasts than Gram-negative bacteria. The extracts exhibited antioxidant and antimicrobial activities in different concentrations. Total phenolic concentration of the extracts was estimated with Folin-Ciocalteu reagent using gallic acid as standard. The total phenolic content varied widely in different parts of the three tested Inula species, ranging from 21.1 +/- 0.8 to 190.9 +/- 6.1 mg GAE/g extract. Phenolic components, such as chlorogenic acid, caffeic acid, rutin, myricetin, quercetin, luteolin and kaempferol were quantified by HPLC-DAD in the methanol extracts of the Inula species. It was obvious that the antioxidant and antimicrobial properties of the plants were due to the phenolics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Gallussäure, 97.5-102.5% (titration)
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Sigma-Aldrich
Chlorogensäure, ≥95% (titration)
Sigma-Aldrich
Antioxidans-Assay-Kit, sufficient for 200 tests
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Rutin Hydrat, ≥94% (HPLC), powder
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline