Merck
  • Insulin/insulin-like growth factor (IGF) stimulation abrogates an association between a deubiquitinating enzyme USP7 and insulin receptor substrates (IRSs) followed by proteasomal degradation of IRSs.
Insulin/insulin-like growth factor (IGF) stimulation abrogates an association between a deubiquitinating enzyme USP7 and insulin receptor substrates (IRSs) followed by proteasomal degradation of IRSs.
Biochemical and biophysical research communications (2012-05-29)
Hidehito Yoshihara, Toshiaki Fukushima, Fumihiko Hakuno, Yasushi Saeki, Keiji Tanaka, Akihiro Ito, Minoru Yoshida, Shun-ichiro Iemura, Tohru Natsume, Tomoichiro Asano, Kazuhiro Chida, Leonard Girnita, Shin-Ichiro Takahashi
ZUSAMMENFASSUNG
Insulin receptor substrates (IRSs) play central roles in insulin/insulin-like growth factor (IGF) signaling and mediate a variety of their bioactivities. IRSs are tyrosine-phosphorylated by activated insulin receptor/IGF-I receptor tyrosine kinase in response to insulin/IGF, and are recognized by signaling molecules possessing the SH2 domain such as phosphatidylinositol 3-kinase (PI3K), leading to the activation of downstream pathways. Recent studies have suggested that degradation of IRSs by the ubiquitin-proteasome pathway leads to impaired insulin/IGF signaling, but the precise mechanism underlying the process is still unclear. In this study, we identified deubiquitinating enzyme ubiquitin specific protease 7 (USP7) as an IRS-2-interacting protein and demonstrated that deubiquitinase activity of USP7 plays important roles in IRS-2 stabilization through the ubiquitin-proteasome pathway. In addition, insulin treatment dissociated USP7 from IRS-2, leading to degradation of IRS-2. This dissociation was prevented by treatment with LY294002, a PI3K inhibitor, indicating that insulin activation of the PI3K pathway leads to dissociation of IRS-2 from USP7 and IRS-2 degradation. We obtained similar results for IRS-1 in cells treated with insulin and for IRS-2 in cells treated with IGF-I. Taken together, this is the first report demonstrating that USP7 is an IRS-1/2 deubiquitinating enzyme forming a negative feedback loop in insulin/IGF signaling.