Merck
  • gamma endorphin, alpha endorphin and Met-enkephalin are formed extracellularly from lipotropin C fragment.
gamma endorphin, alpha endorphin and Met-enkephalin are formed extracellularly from lipotropin C fragment.
Nature (1977-10-13)
B M Austen, D G Smyth, C R Snell
PMID917112