Merck
TYLCV-Is movement in planta does not require V2 protein.
Virology (2015-02-04)
Hagit Hak, Yael Levy, Sam A Chandran, Eduard Belausov, Abraham Loyter, Moshe Lapidot, Yedidya Gafni
ZUSAMMENFASSUNG
Tomato yellow leaf curl virus (TYLCV), a major tomato pathogen causing extensive crop losses, is a whitefly-transmitted geminivirus. V2 mutants of TYLCV-Is and related viruses tend to induce symptomless infection with attenuated viral DNA levels, while accumulating close to wild-type DNA levels in protoplasts, suggesting V2 as a movement protein. The discovery of plant-silencing mechanisms and viral silencing suppressors, V2 included, led us to reconsider V2׳s involvement in viral movement. We studied two mutant versions of the virus, one impaired in V2 silencing-suppression activity, and another carrying a non-translatable V2. While both mutant viruses spread in the infected plant to newly emerged leaves at the same rate as the wild-type virus, their DNA-accumulation levels were tenfold lower than in the wild-type virus. Thus, we suggest that the setback in virus proliferation, previously ascribed to a movement impediment, is due to lack of silencing-suppression activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Spermidin, ≥99% (GC)
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Spermidin, BioReagent, Molecular Biology, suitable for cell culture, ≥98%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Spermidin, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Supelco
Glycerin, analytical standard
Supelco
Spermidin, analytical standard
Sigma-Aldrich
Glycerin, tested according to Ph. Eur., anhydrous