Merck
  • Health hazard assessment of occupationally di-(2-ethylhexyl)-phthalate-exposed workers in China.
Health hazard assessment of occupationally di-(2-ethylhexyl)-phthalate-exposed workers in China.
Chemosphere (2014-06-30)
Wenxin Wang, Xiaobing Xu, Chinbay Q Fan
ZUSAMMENFASSUNG
Di-(2-ethylhexyl)-phthalate (DEHP) is a potential hazard to human health. The effects of occupational high level DEHP exposure on human health were evaluated by measuring the plasma cholinesterase, residues, renal and hepatic biochemical markers. The study was conducted in three representative polyvinyl chloride manufacturing facilities from large size (S1), medium side (S2) to small size (S3). Total 456 adult males including 352 exposed workers (occupational) and 104 control workers (background) were selected. The average DEHP concentrations in respirable particulate matter were 233, 291, and 707 μg m(-3) for S1-S3, respectively, compared with 0.26 μg m(-3) in the background atmosphere (labeled by S4). The results showed significant decreases in post exposure plasma cholinesterase (PChE) levels (<30%) from the exposed workers as compared to baseline. These exposed workers had been evaluated for plasma DEHP residues. Regression analyses explored that PChE decreased significantly with increasing plasma DEHP residues. Serum aspartate aminotransferase, alanine aminotransferase, creatinine, urea, gamma glutamyltransferase, malondialdehyde, total antioxidant and C-reactive protein were significantly raised as compared to the controls. Of the 352 exposed workers, 116 (33.0%) had a daily DEHP intake 22.7 μg kg bw(-1)d(-1) , which is more than 20 μg kg bw(-1)d(-1) specified by the US Environmental Protection Agency. The study demonstrated that occupational phthalate exposure produces health hazards.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Aluminiumoxid, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Aluminiumoxid, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aluminiumoxid, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminiumoxid, nanopowder, <50 nm particle size (TEM)
Supelco
Aceton, analytical standard
Sigma-Aldrich
Aluminiumoxid, powder, 99.99% trace metals basis
Supelco
Hexan, analytical standard
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Aluminiumoxid, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
USP
Aceton, United States Pharmacopeia (USP) Reference Standard
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aluminiumoxid, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminiumoxid, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminiumoxid, Corundum, α-phase, -100 mesh
Supelco
Phthalsäure-bis-2-ethylhexylester, PESTANAL®, analytical standard
Supelco
Aluminiumoxid, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Aluminiumoxid, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminiumoxid, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminiumoxid, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Aluminiumoxid, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminiumoxid, 99.997% trace metals basis