Merck
  • Isolation and characterization of the outer membrane of Escherichia coli with autodisplayed Z-domains.
Isolation and characterization of the outer membrane of Escherichia coli with autodisplayed Z-domains.
Biochimica et biophysica acta (2014-12-22)
Min Park, Gu Yoo, Ji-Hong Bong, Joachim Jose, Min-Jung Kang, Jae-Chul Pyun
ZUSAMMENFASSUNG
"Autodisplay technology" is an expression technique used to display the various recombinant proteins on the outer membrane (OM) of Escherichia coli. The resulting autodisplayed Z-domain has been used to improve the sensitivity of immunoassays. In this work, a facile isolation method of the OM fraction of E. coli with autodisplayed Z-domains was presented using (1) an enzyme reaction for the hydrolysis of the peptidoglycan layer and (2) short centrifugation steps. The purity of the isolated OM fraction was analyzed. For the estimation of contamination with bacterial proteins from other parts of E. coli, Western blots of marker proteins for the OM (OmpA), periplasm (β-lactamase), inner membrane (SecA), and cytoplasm (β-galactosidase) were performed. Additionally, assays of marker components or enzymes from each part of E. coli were carried out including the OM (KDO), inner membrane (NADH oxidase), periplasm (β-lactamase), and cytoplasm (β-galactosidase). The yield of OM isolation using this new method was determined to be 80% of the total OM amount, with less than 1% being contaminants from other parts of E. coli.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adenin, ≥99%
Sigma-Aldrich
2-Nitrophenyl β-D-Galactopyranosid, ≥98% (enzymatic)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Adenin, BioReagent, suitable for cell culture
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥99%
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, Grade AA-1
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 50 mg (per vial)
Sigma-Aldrich
2-Keto-3-Desoxyoctonat Ammoniumsalz, ≥97%
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
Adenin, BioReagent, suitable for plant cell culture, ≥99%
Millipore
Klebsiella-selektiver Zusatz, suitable for microbiology
Supelco
Adenin, Pharmaceutical Secondary Standard; Certified Reference Material
Adenin, European Pharmacopoeia (EP) Reference Standard