Merck
The effects and possible mechanisms of puerarin to treat endometriosis model rats.
Evidence-based complementary and alternative medicine : eCAM (2015-03-31)
Jin Yu, Li Zhao, Danying Zhang, Dongxia Zhai, Wei Shen, Lingling Bai, Yiqun Liu, Zailong Cai, Ji Li, Chaoqin Yu
ZUSAMMENFASSUNG
Objective. To explore the effects of puerarin to treat endometriosis (EMT) model rats and the possible regulatory mechanisms. Methods. EMT model rats were surgically induced by autotransplantion of endometrial tissues. The appropriate dosage of puerarin to treat EMT model rats was determined by observing the pathologic morphology of ectopic endometrial tissues and by detecting the levels of estradiol (E2) and prostaglandin E2 (PGE2) of both serum and ectopic endometrial tissues. The related genes and proteins of ectopic endometrial tissues were analyzed by Real-time PCR and immunohistochemistry (IHC) to explore the possible mechanisms. Results. Puerarin could reduce the levels of E2 and PGE2 and prevent the growth of ectopic endometrium tissues by inhibiting the expression of aromatase cytochrome P450 (p450arom) and cyclooxygenase-2 (cox-2); puerarin could adjust the anabolism of E2 by upregulating the expression of 17β-hydroxysteroid-2 (17β-hsd-2) and downregulating the expression of 17β-hydroxysteroid-1 (17β-hsd-1) of the ectopic endometrium tissues; puerarin could increase the expression of ERβ and improve the inflammatory microenvironment of EMT model rats. Conclusions. Our data suggest that puerarin has a therapeutic effect on EMT model rats and could be a potential therapeutic agent for the treatment of EMT in clinic.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Puerarin, analytical standard