Merck
Effect of the allelic variant of alcohol dehydrogenase ADH1B*2 on ethanol metabolism.
Alcoholism, clinical and experimental research (2014-05-07)
Gaeun Kang, Kyung-Yeol Bae, Sung-Wan Kim, Jin Kim, Hee-Young Shin, Jae-Min Kim, Il-Seon Shin, Jin-Sang Yoon, Jong-Keun Kim
ZUSAMMENFASSUNG
It has been known that ADH1B*2 allele has a protective effect against the development of alcohol dependence. However, the protection mechanism is still unknown. We investigated whether ADH1B gene polymorphism affects ethanol (EtOH) metabolism. In a parent study, we conducted a randomized crossover trials on 24 healthy male subjects who were selected by genotyping: 12 with ALDH2*1/*1 (active form) and 12 with ALDH2*1/*2 (inactive form). In the present study, the 24 subjects were reclassified into 2 groups of 11 with ADH1B*1/*2 and 13 with ADH1B*2/*2 according to the ADH1B genotypes. Each subject was administered 1 of 3 doses of EtOH (0.25, 0.5, 0.75 g/kg) or a placebo in 4 trials. After the administration of alcohol, blood EtOH and acetaldehyde concentrations were measured 9 times over 4 hours. In the case of EtOH, the area under the concentration-time curve from 0 to 4 hours (AUC0-4 ) and the peak blood concentration of EtOH (Cmax ) in subjects with ADH1B*2/*2 were significantly higher than those in subjects with ADH1B*1/*2 at all 3 dosages before stratifying by ALDH2 genotype. However, after stratifying by ALDH2 genotype, a statistically significant difference between ADH1B*2/*2 and ADH1B*1/*2 was found only at the 0.5 g/kg dosage regardless of ALDH2 genotype. In the case of acetaldehyde, the AUC0-4 and Cmax of acetaldehyde of ADH1B*2/*2 after administration of 0.25 g/kg alcohol and the AUC0-4 of acetaldehyde of ADH1B*2/*2 at 0.5 g/kg were significantly higher than corresponding values of ADH1B*1/*2 only in the group of ALDH2*1/*2. Our findings indicate that the blood EtOH concentrations of ADH1B*2/*2 group are higher than those of ADH1B*1/*2 group regardless of ALDH2 genotype, and the blood acetaldehyde concentrations of ADH1B*2/*2 are also higher than those of ADH1B*1/*2 only in the ALDH2*1/*2 group. To our knowledge, this is the first report to demonstrate the association of ADH1B*2 allele with blood EtOH and acetaldehyde levels in humans, and these results suggest that higher blood EtOH and acetaldehyde concentrations in ADH1B*2/*2 may constitute the mechanism of protection against alcoholism by ADH1B*2/*2.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetaldehyd, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyd, natural, FG
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Acetaldehyd, puriss. p.a., anhydrous, ≥99.5% (GC)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Acetaldehyd, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyd, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyd -Lösung, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyd -Lösung, 40 wt. % in H2O
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Acetaldehyd -Lösung, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyd -Lösung, 5 M in THF
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetaldehyd -Lösung, 40 wt. % in isopropanol