Merck
  • HPLC-DAD and UV-spectrophotometry for the determination of lychnopholide in nanocapsule dosage form: validation and application to release kinetic study.
HPLC-DAD and UV-spectrophotometry for the determination of lychnopholide in nanocapsule dosage form: validation and application to release kinetic study.
Journal of chromatographic science (2012-12-19)
Renata Tupinambá Branquinho, Vanessa Carla Furtado Mosqueira, Eunice Kazue Kano, Jacqueline de Souza, Diego Dias Ramos Dorim, Dênia Antunes Saúde-Guimarães, Marta de Lana
ZUSAMMENFASSUNG
Simple and sensitive methods using high-performance liquid chromatography-diode array detection (HPLC-DAD) and ultraviolet (UV)-spectrophotometry were developed and compared to quantify lychnopholide (LYC) in poly-ε-caprolactone nanocapsules and to study its release kinetics. Both methods were validated concerning their specificity, linearity, limits of detection and quantification, precision, accuracy and stability. HPLC-DAD analyses were conducted using an RP C18 column, isocratic elution with a methanol-water (60:40 v/v) mobile phase at 0.8 mL/min flow rate and detection at 265 nm. The linear response (r(2) > 0.999) was obtained within a concentration range of 2-25 µg/mL using HPLC-DAD and 5-40 µg/mL using spectrophotometry. Intra-day and inter-day precision were obtained with low relative standard deviation values. The accuracy of the methods was within the range 98-101% for HPLC-DAD and from 96-100% for UV-spectrophotometry. Both methods were suitable to be applied for the determination of drug loading percentage (>96%) and encapsulation efficiency (>90%). Furthermore, the sensitivity of HPLC-DAD method allows studies of LYC release/dissolution in sink conditions. LYC presented 100% dissolution after 24 h, whereas only 60% of LYC was released from the nanocapsule dosage form, with no burst effect. The methods fulfilled all validation parameters evaluated for LYC quantification in the polymeric nanocapsules and have proven to be accurate, selective and sensitive in the previously mentioned applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
p-Anisaldehyd, 98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Hexan, anhydrous, 95%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Aceton, analytical standard
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Ethylacetat, analytical standard
Supelco
Hexan, analytical standard