Merck
  • Human Schistosoma haematobium antifecundity immunity is dependent on transmission intensity and associated with immunoglobulin G1 to worm-derived antigens.
Human Schistosoma haematobium antifecundity immunity is dependent on transmission intensity and associated with immunoglobulin G1 to worm-derived antigens.
The Journal of infectious diseases (2014-07-09)
Shona Wilson, Frances M Jones, Govert J van Dam, Paul L A M Corstjens, Gilles Riveau, Colin M Fitzsimmons, Moussa Sacko, Birgitte J Vennervald, David W Dunne
ZUSAMMENFASSUNG
Immunity that reduces worm fecundity and, in turn, reduces morbidity is proposed for Schistosoma haematobium, a parasite of major public health importance. Mathematical models of epidemiological trends suggest that antifecundity immunity is dependent on antibody responses to adult-worm-derived antigen. For a Malian cohort (age, 5-29 years) residing in high-transmission fishing villages or a moderate-transmission village, worm fecundity was assessed using the ratio of urinary egg excretion to levels of circulating anodic antigen, a Schistosoma-specific antigen that is steadily secreted by adult worms. Fecundity was modeled against host age, infection transmission intensity, and antibody responses specific to soluble worm antigen (SWA), tegument allergen-like 1, and 28-kDa glutathione-S-transferase. Worm fecundity declined steadily until a host age of 11 years. Among children, host age and transmission were negatively associated with worm fecundity. A significant interaction term between host age and transmission indicates that antifecundity immunity develops earlier in high-transmission areas. SWA immunoglobulin G1 (IgG1) levels explained the effect of transmission on antifecundity immunity. Antifecundity immunity, which is likely to be protective against severe morbidity, develops rapidly during childhood. Antifecundity immunity is associated with SWA-IgG1, with higher infection transmission increasing this response at an earlier age, leading to earlier development of antifecundity immunity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
o-Phenylendiamin, flaked, 99.5%
Sigma-Aldrich
1,3-Diaminopropan, ≥99%
Sigma-Aldrich
Tributylphosphat, ≥99%
Sigma-Aldrich
Tributylphosphat, 97%
Sigma-Aldrich
o-Phenylendiamin, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylendiamin, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylendiamin, sublimed, ≥99%
Supelco
Tributylphosphat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tributylphosphat, puriss., ≥99.0% (GC)
Tri-n-butyl-phosphat, European Pharmacopoeia (EP) Reference Standard
Supelco
Tributylphosphat, for extraction analysis, ≥99.0% (GC)