Merck
  • Development and application of a microfluidic in-situ analyzer for dissolved Fe and Mn in natural waters.
Development and application of a microfluidic in-situ analyzer for dissolved Fe and Mn in natural waters.
Talanta (2015-02-24)
Ambra Milani, Peter J Statham, Matt C Mowlem, Douglas P Connelly
ZUSAMMENFASSUNG
The redox sensitive trace metals iron and manganese are two important elements that help shape the biogeochemistry of aquatic systems and thus their measurement is important. Current laboratory methods are expensive, time consuming and cannot provide the spatial and temporal resolution needed to characterize these elements in natural waters. Here we describe the first autonomous analyzer capable of providing vertical profiles as well as routine in-situ determinations of dissolved Fe(II) and Mn in aquatic environments. The spectrophotometric sensor uses microfluidic methods (Lab-on-a-chip technology) and mixes reagents and samples using a novel in-cell diffusion process. Fe(II) and Mn can be measured with a frequency of up to 12 and 6 samples per hour respectively with limits of detection of 27nM for Fe(II), 2.1% precision (n=20), and 28nM for Mn, 2.4% precision (n=19). The device combines relatively low cost, low power usage, low reagent consumption, portability, and tolerance to pressures up to at least 170 bars, with high precision and accuracy. We present data from a successful demonstration of the sensor during a cruise to the Gotland and Landsort Deep Basins of the Baltic Sea.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Natriumacetat, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Borsäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Natriumacetat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Borsäure, ReagentPlus®, ≥99.5%
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
Borsäure, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumacetat, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
USP
Ascorbinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Borsäure, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%