Merck
T cells and lung injury. Impact of rapamycin.
American journal of respiratory cell and molecular biology (2014-03-20)
Takeshi Nakajima, Ko-Wei Lin, Jinghong Li, Halvor S McGee, Jennifer M Kwan, David L Perkins, Patricia W Finn
ZUSAMMENFASSUNG
Acute lung injury (ALI) is characterized by pulmonary inflammation and edema. Innate immune cells (e.g., neutrophils and macrophages) are major contributors to inflammation in ALI. Less is known regarding the role of T cells. We examined the effects of rapamycin on inflammation in a LPS-induced murine model of ALI. Rapamycin was administered before and after initiation of injury. Inflammatory parameters, including bronchoalveolar lavage cell counts, T cell surface markers (i.e., cytotoxic T lymphocyte antigen 4 [CTLA4] and fork head-winged helix transcription factor [Foxp3]), T cell activation (CD69), IL-6, and IL-10 were analyzed. Rapamycin significantly decreased inflammatory parameters and decreased Foxp3, CTLA4, and CD69 in CD4(+) T cells. Rapamycin administration before or after the onset of lung injury, as well as systemically or by pulmonary routes, ameliorates inflammation in ALI.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumfluorid, ACS reagent, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Natriumpyrophosphat, ≥95%
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumfluorid, ReagentPlus®, ≥99%
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumfluorid, 99.99% trace metals basis
Supelco
Natriumfluorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Okadasäure aus Prorocentrum concavum, 92-100% (HPLC)
Sigma-Aldrich
Natriumfluorid, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Natriumfluorid, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumfluorid, BioXtra, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Natriumfluoridlösung
Sigma-Aldrich
Natriumfluorid, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumfluorid 0,5 M -Lösung
Supelco
Fluorid-Ionenlösung für ISE, 0.1 M F-, for ion-selective electrodes