Merck
Intranasal triamcinolone and growth velocity.
Pediatrics (2015-01-28)
David P Skoner, William E Berger, Sandra M Gawchik, Akbar Akbary, Chunfu Qiu
ZUSAMMENFASSUNG
Inadequate designs and conflicting results from previous studies prompted the US Food and Drug Administration to publish guidelines for the design of clinical trials evaluating the effects of orally inhaled and intranasal corticosteroids on the growth of children. This study conformed to these guidelines to evaluate the effect of triamcinolone acetonide aqueous nasal spray (TAA-AQ) on the growth of children with perennial allergic rhinitis (PAR). This randomized, double-blind, placebo-controlled, parallel-group, multicenter study evaluated the effect of once-daily TAA-AQ (110 μg) on the growth velocity (GV) of children aged 3-9 years with PAR by using stadiometry at baseline (4-6 months), during treatment (12 months), and at follow-up (2 months). Hypothalamus-pituitary-adrenal (HPA) axis function was assessed by measuring urinary cortisol levels. Details of adverse events were recorded. Of 1078 subjects screened, 299 were randomized, and 216 completed the study (placebo, 107; TAA-AQ, 109). In the primary analysis (modified intent-to-treat: placebo, 133; TAA-AQ, 134), least-squares mean GV during treatment was lower in the TAA-AQ group (5.65 cm/year) versus placebo (6.09 cm/year). The difference (-0.45 cm/year; 95% confidence interval: -0.78 to -0.11; P = .01), although clinically nonsignificant, was evident within 2 months of treatment and stabilized thereafter. At follow-up, the GV approached baseline (6.70 cm/year) in the TAA-AQ group (6.59 cm/year) and decreased slightly in the placebo group (5.89 cm/year vs 6.06 cm/year at baseline). No HPA axis suppression was observed. By using rigorous Food and Drug Administration-recommended design elements, this study detected a small, statistically significant effect of TAA-AQ on the GV of children with PAR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Supelco
Hydrocortison, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydrocortison, United States Pharmacopeia (USP) Reference Standard
Supelco
Kreatinin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortison, meets USP testing specifications
Hydrocortison für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Hydrocortison, European Pharmacopoeia (EP) Reference Standard
Hydrocortison, British Pharmacopoeia (BP) Assay Standard