Merck
  • Determination of quaternary ammonium compounds in oranges and cucumbers using QuEChERS extraction and ultra-performance liquid chromatography/tandem mass spectrometry.
Determination of quaternary ammonium compounds in oranges and cucumbers using QuEChERS extraction and ultra-performance liquid chromatography/tandem mass spectrometry.
Journal of AOAC International (2014-08-26)
Francisco Javier Arrebola-Liébanas, María Angeles Herrera Abdo, José Luis Fernandez Moreno, José L Martínez-Vidal, Antonia Garrido Frenich
ZUSAMMENFASSUNG
A simple and fast method has been developed for determining relevant quaternary ammonium compounds in cucumber and orange samples. The target compounds were benzoalkonium chloride (BAC-10, BAC-12, BAC-14, and BAC-16), didecyldimethylammonium chloride, and benzethonium chloride, all frequently used biocides in the agrifood industry. An extraction based on the buffered Quick, Easy, Cheap, Effective, Rugged, and Safe method and determination by ultra-performance LC/MS/MS that eluted the biocides in less than 5 min were used. The method was fully validated and implemented in a UNE-EN-ISO/IEC 17025 accredited laboratory for its application to the analysis of real samples. Performance characteristics of the method are reported, including an estimation of measurement uncertainty. Calibration curves were set between 0.01 and 0.150 mg/kg, LOD values were always between 0.4 and 1.0 microg/kg, LOQ values were in the range 1-4 microg/kg, recovery was between 81 and 115%, intraday and interday precision were always lower than 17% (expressed as RSD), and expanded uncertainty was always lower than 40%. The validation was accomplished for the two studied matrixes at spiking concentrations of 0.011 and 0.050 mg/kg. The method has been applied to the analysis of 30 cucumber and orange samples that were found to contain concentrations of BAC-12 that ranged between 0.015 and 0.210 mg/kg and of BAC-14 between 0.018 and 0.081 mg/kg.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hexadecyltrimethylammoniumchlorid -Lösung, 25 wt. % in H2O
Sigma-Aldrich
Hexadecyl-trimethylammoniumchlorid, ≥98.0% (NT)
Sigma-Aldrich
Benzyl-dimethyl-hexadecylammoniumchlorid, cationic detergent
Sigma-Aldrich
Benzyldimethyldodecylammoniumchlorid, ≥99.0% (AT)