Merck
  • Detection of benzalkonium chloride resistance in community environmental isolates of staphylococci.
Detection of benzalkonium chloride resistance in community environmental isolates of staphylococci.
Journal of medical microbiology (2014-03-04)
Gui-Xin He, Michael Landry, Huizhong Chen, Conner Thorpe, Dennis Walsh, Manuel F Varela, Hongmiao Pan
ZUSAMMENFASSUNG
We isolated a total of 653 strains from 64 community environmental samples in Massachusetts, USA. Among these isolates, 9.65 % (63 strains) were benzalkonium chloride (BC)-resistant staphylococci. All BC-resistant strains were collected from surfaces upon which antibacterial wipes or antibacterial sprays containing 0.02-0.12 % BC had frequently been used in the fitness centres. However, isolates from surfaces upon which antibacterial wipes or antibacterial sprays had not been used were all sensitive to BC. All BC-resistant strains were also resistant to erythromycin, penicillin and ampicillin. In addition, 51 strains showed resistance to cetyltrimethylammonium bromide (CTAB), 15 strains showed resistance to chloramphenicol, 12 strains showed resistance to ciprofloxacin and four strains showed resistance to meticillin. Resistance gene analysis demonstrated that 41 strains contained qacA/B, 30 strains had qacC, 25 strains contained qacG, 16 strains had qacH and eight strains contained qacJ. These data indicate that application of BC is associated with environmental staphylococcal antimicrobial resistance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Supelco
D-Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Mannitol, European Pharmacopoeia (EP) Reference Standard
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination