Merck
  • Polymeric micelles loaded with platinum anticancer drugs target preangiogenic micrometastatic niches associated with inflammation.
Polymeric micelles loaded with platinum anticancer drugs target preangiogenic micrometastatic niches associated with inflammation.
Journal of controlled release : official journal of the Controlled Release Society (2014-06-24)
Hailiang Wu, Horacio Cabral, Kazuko Toh, Peng Mi, Yi-Chun Chen, Yu Matsumoto, Naoki Yamada, Xueying Liu, Hiroaki Kinoh, Yutaka Miura, Mitsunobu R Kano, Hiroshi Nishihara, Nobuhiro Nishiyama, Kazunori Kataoka
ZUSAMMENFASSUNG
Nanocarriers have been used for specific delivery of therapeutic agents to solid tumors based on the enhanced permeability and retention in cancerous tissues. Despite metastasis is the main reason of cancer-related death and a priority for nanocarrier-based therapies, the targeting ability of nanocarriers to the metastatic disease is poorly understood, especially for preangiogenic micrometastases as nanocarriers usually use the malignant neovasculature for enhancing their accumulation. Thus, herein, we studied the ability of micellar nanocarriers incorporating (1,2-diaminocyclohexane)platinum(II) (DACHPt) for treating liver metastases of bioluminescent murine colon adenocarcinoma C-26, during overt and preangiogenic metastatic stages. After intravenous injection, DACHPt-loaded micelles (DACHPt/m) effectively inhibited the tumor growth in both metastatic tumor models. While the anticancer activity of the micelles against overt metastases was associated with their selective accumulation in cancerous tissues having neovasculature, the ability of DACHPt/m to target preangiogenic metastases was correlated with the inflammatory microenvironment of the niche. This targeting capability of polymeric micelles to preangiogenic metastasis may provide a novel approach for early diagnosis and treatment of metastases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-Aktin-Antikörper , α-Glattmuskel, clone 1A4, ascites fluid
Sigma-Aldrich
Bis(trichlormethyl)carbonat, reagent grade, 98%
Sigma-Aldrich
Oxaliplatin, powder
Sigma-Aldrich
Bis(trichlormethyl)carbonat, purum, ≥99.0% (AT)
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard
Supelco
Oxaliplatin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Oxaliplatin, United States Pharmacopeia (USP) Reference Standard
Isofluran, European Pharmacopoeia (EP) Reference Standard
Celecoxib, European Pharmacopoeia (EP) Reference Standard