Merck
  • SOX2 oncogenes amplified and operate to activate AKT signaling in gastric cancer and predict immunotherapy responsiveness.
SOX2 oncogenes amplified and operate to activate AKT signaling in gastric cancer and predict immunotherapy responsiveness.
Journal of cancer research and clinical oncology (2014-04-23)
Yajun Tian, Xin Jia, Shengxiang Wang, Yongsheng Li, Peng Zhao, Da Cai, Zequan Zhou, Junmin Wang, Yi Luo, Maosheng Dong
ZUSAMMENFASSUNG
Gastric cancer is the second leading cause of cancer mortality in the world. Whether the oncogene, amplified on chromosome 3q26, SOX2, a master transcriptional regulator of stemness, operate to drive strong growth phenotype in gastric cancer were unknown. The gene expression changes of SOX2 in human gastric cancer tissues compared with non-cancerous tissues was detected using real-time quantitative reverse transcriptase-polymerase chain reaction (QRT-PCR) analysis and immunohistochemistry, which identified the gene overexpression of SOX2 in gastric cancer. Moreover, we discovered that SOX2 promoted cancer cell proliferation in vitro/vivo and SOX2 expression correlated with elevated AKT phosphorylation in gastric cancer, while the AKT phosphorylation was required for SOX2's oncogenic effects. Next, our data point to the usefulness of SOX2 overexpression, as a new predictive marker for responsiveness to trastuzumab. SOX2 is a commonly activated tumor promoter that activate AKT signaling in gastric cancer and a new predictive marker for targeted therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
cis-Diamminplatin(II)-dichlorid, crystalline
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diaminplatin(II)-dichlorid, ≥99.9% trace metals basis
Sigma-Aldrich
trans-Platin(II)diammindichlorid
Cisplatin Unreinheit A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard