Merck
  • Encapsulation of folic acid in food hydrocolloids through nanospray drying and electrospraying for nutraceutical applications.
Encapsulation of folic acid in food hydrocolloids through nanospray drying and electrospraying for nutraceutical applications.
Food chemistry (2014-08-31)
Rocío Pérez-Masiá, Rubén López-Nicolás, Maria Jesús Periago, Gaspar Ros, Jose M Lagaron, Amparo López-Rubio
ZUSAMMENFASSUNG
In this work, two different technologies (electrospraying and nanospray drying) were evaluated for the encapsulation of folic acid using both a whey protein concentrate (WPC) matrix and a commercial resistant starch. The morphology of the capsules, molecular organization of the matrices upon encapsulation, encapsulation efficiency, and stability of the folic acid within the capsules under different storage conditions and upon thermal exposure were studied. Results showed that spherical nano-, submicro- and microcapsules were obtained through both techniques, although electrospraying led to smaller capsule sizes and to an enhanced control over their size distribution. Greater encapsulation efficiency was observed using WPC as encapsulating matrix, probably related to interactions between the protein and folic acid which favoured the incorporation of the bioactive. The best results in terms of bioactive stabilization in the different conditions assayed were also obtained for the WPC capsules, although both materials and encapsulation techniques led to improved folic acid stability, especially under dry conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Folsäure, ≥97%
Sigma-Aldrich
Folsäure, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Supelco
Folsäure, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Folsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Folsäure, meets USP testing specifications
Folsäure, European Pharmacopoeia (EP) Reference Standard