Merck
Role of cannabinoid CB2 receptor in the reinforcing actions of ethanol.
Addiction biology (2013-07-17)
Antonio Ortega-Álvaro, Alexander Ternianov, Auxiliadora Aracil-Fernández, Francisco Navarrete, Maria Salud García-Gutiérrez, Jorge Manzanares
ZUSAMMENFASSUNG
This study examines the role of the cannabinoid CB2 receptor (CB2 r) on the vulnerability to ethanol consumption. The time-related and dose-response effects of ethanol on rectal temperature, handling-induced convulsions (HIC) and blood ethanol concentrations were evaluated in CB2 KO and wild-type (WT) mice. The reinforcing properties of ethanol were evaluated in conditioned place preference (CPP), preference and voluntary ethanol consumption and oral ethanol self-administration. Water-maintained behavior schedule was performed to evaluate the degree of motivation induced by a natural stimulus. Preference for non-alcohol tastants assay was performed to evaluate the differences in taste sensitivity. Tyrosine hydroxylase (TH) and μ-opioid receptor gene expressions were also measured in the ventral tegmental area and nucleus accumbens (NAcc), respectively. CB2 KO mice presented increased HIC score, ethanol-CPP, voluntary ethanol consumption and preference, acquisition of ethanol self-administration, and increased motivation to drink ethanol compared with WT mice. No differences were found between genotypes in the water-maintained behavior schedule or preference for non-alcohol tastants. Naïve CB2 KO mice presented increased μ-opioid receptor gene expression in NAcc. Acute ethanol administration (1-2 g/kg) increased TH and μ-opioid receptor gene expressions in CB2 KO mice, whereas the lower dose of ethanol decreased TH gene expression in WT mice. These results suggest that deletion of the CB2 r gene increased preference for and vulnerability to ethanol consumption, at least in part, by increased ethanol-induced sensitivity of the TH and μ-opioid receptor gene expressions in mesolimbic neurons. Future studies will determine the role of CB2 r as a target for the treatment of problems related with alcohol consumption.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Mettler-Toledo-Kalibriersubstanz ME 51143091, Saccharin, traceable to primary standards (LGC)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharin, United States Pharmacopeia (USP) Reference Standard
Saccharin, European Pharmacopoeia (EP) Reference Standard