Merck
  • Regulation of postsynaptic plasticity genes' expression and topography by sustained dopamine perturbation and modulation by acute memantine: relevance to schizophrenia.
Regulation of postsynaptic plasticity genes' expression and topography by sustained dopamine perturbation and modulation by acute memantine: relevance to schizophrenia.
Progress in neuro-psychopharmacology & biological psychiatry (2014-07-16)
Felice Iasevoli, Elisabetta F Buonaguro, Chiara Sarappa, Federica Marmo, Gianmarco Latte, Rodolfo Rossi, Anna Eramo, Carmine Tomasetti, Andrea de Bartolomeis
ZUSAMMENFASSUNG
A relevant role for dopamine-glutamate interaction has been reported in the pathophysiology and treatment of psychoses. Dopamine and glutamate may interact at multiple levels, including the glutamatergic postsynaptic density (PSD), an electron-dense thickening that has gained recent attention as a switchboard of dopamine-glutamate interactions and for its role in synaptic plasticity. Recently, glutamate-based strategies, such as memantine add-on to antipsychotics, have been proposed for refractory symptoms of schizophrenia, e.g. cognitive impairment. Both antipsychotics and memantine regulate PSD transcripts but sparse information is available on memantine's effects under dopamine perturbation. We tested gene expression changes of the Homer1 and PSD-95 PSD proteins in models of sustained dopamine perturbation, i.e. subchronic treatment by: a) GBR-12909, a dopamine receptor indirect agonist; b) haloperidol, a D2R antagonist; c) SCH-23390, a dopamine D1 receptor (D1R) antagonist; and d) SCH-23390+haloperidol. On the last day of treatment, rats were acutely treated with vehicle or memantine. The Homer1a immediate-early gene was significantly induced by haloperidol and by haloperidol+SCH-23390. The gene was not induced by SCH-23390 per se or by GBR-12909. Expression of the constitutive genes Homer1b/c and PSD-95 was less affected by these dopaminergic paradigms. Acute memantine administration significantly increased Homer1a expression by the dopaminergic compounds used herein. Both haloperidol and haloperidol+SCH-23390 shifted Homer1a/Homer1b/c ratio of expression toward Homer1a. This pattern was sharpened by acute memantine. Dopaminergic compounds and acute memantine also differentially affected topographic distribution of gene expression and coordinated expression of Homer1a among cortical-subcortical regions. These results indicate that dopaminergic perturbations may affect glutamatergic signaling in different directions. Memantine may help partially revert dopamine-mediated glutamatergic dysfunctions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Memantin -hydrochlorid, ≥98% (GC)