Merck
  • Regeneration of corneal epithelium utilizing a collagen vitrigel membrane in rabbit models for corneal stromal wound and limbal stem cell deficiency.
Regeneration of corneal epithelium utilizing a collagen vitrigel membrane in rabbit models for corneal stromal wound and limbal stem cell deficiency.
Acta ophthalmologica (2014-12-17)
J Jeremy Chae, Winnette McIntosh Ambrose, Freddy A Espinoza, Daniel G Mulreany, Shengyong Ng, Toshiaki Takezawa, Morgana M Trexler, Oliver D Schein, Roy S Chuck, Jennifer H Elisseeff
ZUSAMMENFASSUNG
This study was performed to evaluate the potential of a collagen-based membrane, collagen vitrigel (CV), for reconstructing corneal epithelium in the stromal wound and limbal stem cell deficiency (LSCD) models. Three groups of rabbits were used in the stromal wound model: CV affixed using fibrin glue (CV + FG group, n = 9), fibrin glue only (FG group, n = 3) and an untreated control group (n = 3). In the LSCD model, one group received CV containing human limbal epithelial cells (CV + hLEC group, n = 2) and the other was an untreated control (n = 1). Gross observation, including fluorescent staining, pathological examination, immunohistochemistry and electron microscopy, was used to evaluate the effect of CV on the corneal epithelium. In the stromal wound model, fluorescent staining showed that epithelial reconstruction occurred as rapidly in the CV + FG group as it did in the control group. The pathological examination proved that the CV supported a healthy corneal epithelium in the CV + FG group, whereas FG led to hypertrophy and inappropriate differentiation of corneal epithelium in the FG group. In the LSCD model, the corneas in the CV + hLEC group showed sustained tissue transparency with good epithelialization, low inflammatory response and reduced neovascularization. However, the control cornea was translucent and showed high amounts of inflammation and neovascularization. We have demonstrated that CV supports corneal epithelial differentiation and prevents epithelial hypertrophy, in addition to serving as a scaffold for hLEC transplantation, without complications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Hexamethyldisilazan, reagent grade, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Osmiumtetroxid, ReagentPlus®, 99.8%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Osmiumtetroxid -Lösung, 4 wt. % in H2O
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, ACS reagent
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hexamethyldisilazan, ReagentPlus®, 99.9%
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)