Merck
  • Modification to AOAC official methods 2009.01 and 2011.25 to allow for minor overestimation of low molecular weight soluble dietary fiber in samples containing starch.
Modification to AOAC official methods 2009.01 and 2011.25 to allow for minor overestimation of low molecular weight soluble dietary fiber in samples containing starch.
Journal of AOAC International (2014-07-24)
Barry V Mccleary
ZUSAMMENFASSUNG
AOAC Official Methods 2009.01 and 2011.25 have been modified to allow removal of resistant maltodextrins produced on hydrolysis of various starches by the combination of pancreatic alpha-amylase and amyloglucosidase (AMG) used in these assay procedures. The major resistant maltodextrin, 6(3),6(5)-di-alpha-D-glucosyl maltopentaose, is highly resistant to hydrolysis by microbial alpha-glucosidases, isoamylase, pullulanase, pancreatic, bacterial and fungal alpha-amylase and AMG. However, this oligosaccharide is hydrolyzed by the mucosal alpha-glucosidase complex of the pig small intestine (which is similar to the human small intestine), and thus must be removed in the analytical procedure. Hydrolysis of these oligosaccharides has been by incubation with a high concentration of a purified AMG at 60 degrees C. This incubation results in no hydrolysis or loss of other resistant oligosaccharides such as FOS, GOS, XOS, resistant maltodextrins (e.g., Fibersol 2) or polydextrose. The effect of this additional incubation with AMG on the measured level of low molecular weight soluble dietary fiber (SDFS) and of total dietary fiber in a broad range of samples is reported. Results from this study demonstrate that the proposed modification can be used with confidence in the measurement of dietary fiber.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Supelco
Natronlauge, 49-51% in water, eluent for IC
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Supelco
Natronlauge-Konzentrat, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Supelco
Essigsäure, analytical standard
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG