Merck
  • Swelling, diffusion, network parameters and adsorption properties of IPN hydrogel of chitosan and acrylic copolymer.
Swelling, diffusion, network parameters and adsorption properties of IPN hydrogel of chitosan and acrylic copolymer.
Materials science & engineering. C, Materials for biological applications (2014-10-05)
Bidyadhar Mandal, Samit Kumar Ray
ZUSAMMENFASSUNG
Interpenetrating network (IPN) type hydrogels of a biopolymer and a synthetic polymer were prepared from chitosan and crosslink copolymer of acrylic acid, sodium acrylate and hydroxyethyl methacrylate. Acrylic acid, sodium acrylate, hydroxyethyl methacrylate and N'N'-methylenebisacrylamide (MBA) monomers were free radically copolymerized and crosslinked in aqueous solution of chitosan. Several IPN hydrogels were prepared by varying concentrations of initiator, crosslinker (MBA) and weight% of chitosan . These hydrogels were characterized by free acid content, pH at point of zero charge (PZC), FTIR, DTA-TGA, SEM and XRD. The swelling and diffusion characteristics, network parameters and adsorption of cationic methyl violet (MV) and anionic congo red (CR) dyes by these hydrogels were studied. The hydrogels showed high adsorption (9.5-119 mg/g for CR and 9.2-98 mg/g for MV) and removal% (98-73% for CR and 94-66% for MV) over the feed concentration of 10-140 mg/l dye in water. The isotherms and kinetics of dye adsorption by the hydrogels were also studied.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumbromid, ≥99% trace metals basis
Sigma-Aldrich
Ammoniumpersulfat, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammoniumpersulfat, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Acrylsäure, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Kaliumbromid, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammoniumpersulfat, reagent grade, 98%
Sigma-Aldrich
Natriummetabisulfit, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Hydroxyethylmethacrylat, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
2-Hydroxyethylmethacrylat, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
Natriummetabisulfit, reagent grade, 97%
Sigma-Aldrich
Natriummetabisulfit, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5%
Sigma-Aldrich
Kaliumbromid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Ammoniumpersulfat, BioUltra, ≥98.0% (RT)
Supelco
Natriummetabisulfit, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kaliumbromid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Kaliumbromid, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ammoniumpersulfat, BioXtra, ≥98.0%
Sigma-Aldrich
Kaliumbromid, anhydrous, powder, 99.999% trace metals basis
Supelco
Natriummetabisulfit, analytical standard
Sigma-Aldrich
Kaliumbromid, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Kaliumbromid, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Hydroxyethylmethacrylat
Sigma-Aldrich
Kaliumbromid, BioXtra, ≥99.0%
USP
Natriummetabisulfit, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriummetabisulfit, tested according to Ph. Eur.
Sigma-Aldrich
Ammoniumpersulfat, free-flowing, Redi-Dri, reagent grade, 98%