Merck
Biofilm community diversity after exposure to 0·4% stannous fluoride gels.
Journal of applied microbiology (2014-09-30)
C Reilly, K Rasmussen, T Selberg, J Stevens, R S Jones
ZUSAMMENFASSUNG
To test the effect of 0·4% stannous fluoride (SnF2 ) glycerine-based gels on specific portions of the bacterial community in both a clinical observational study and in vitro multispecies plaque-derived (MSPD) biofilm model. Potential changes to specific portions of the bacterial community were determined through the Human Oral Microbial Identification Microarray (HOMIM). Both the observational clinical study and the biofilm model showed that short-term use of 0·4% SnF2 gel has little effect on the bacterial community depicted by hierarchical cluster analysis. The amount of plaque accumulation on a subject's teeth, which was measured by plaque index scores, failed to show statistical significant changes over the two baselines or after treatment (P = 0·9928). The in vitro results were similar when examining the effect of 0·4% SnF2 gels on biofilm adherence through a crystal violet assay (P = 0·1157). The bacteria within the dental biofilms showed resilience in maintaining the overall community diversity after exposure to 0·4% SnF2 topical gels. The study supports that the immediate benefits of using 0·4% SnF2 gels in children may be strictly from fluoride ions inhibiting tooth demineralization rather than delivering substantial antimicrobial effects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Hydroxylapatit, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Hydroxyapatit, powder, synthetic
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Zinn(II)-fluorid, 99%
Supelco
Glycerin, analytical standard
Sigma-Aldrich
Calciumphosphat tribasisch, 34.0-40.0% Ca basis
Sigma-Aldrich
Hydroxyapatit, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Hydroxylapatit, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxylapatit, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Glycerin, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Hydroxylapatit, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxylapatit, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Calciumphosphat tribasisch, BioReagent, suitable for plant cell culture, powder