Merck
The influence of the gastrointestinal tract and the liver on cystatin C serum concentrations.
Scandinavian journal of clinical and laboratory investigation (2014-04-17)
Erland J Erlandsen, Jan Abrahamsen
ZUSAMMENFASSUNG
The purpose of this study in humans was to examine the influence of the gastrointestinal tract and liver on the serum concentrations of cystatin C. Eighteen healthy volunteers and 28 patients suspected of having chronic intestinal ischemia underwent catheterization of the abdominal aorta and the central hepatic vein. Blood samples were taken simultaneously from the abdominal aorta and the central hepatic vein 60, 90 and 120 minutes after the start of the investigation. After the first blood sample, a standard liquid meal was ingested. Measurement of splanchnic blood flow was performed using the Fick principle with constant infusion of (99m)Tc-Bridatec. Angiography was performed at the end of the investigation. The splanchnic blood flow increased significantly postprandially in the healthy volunteers and in the patients with normal angiography by 0.613-0.698 L/min and increased non- significantly in the patients with abnormal angiography (n = 5) by 0.135 L/min on average. ANOVA and the Bonferroni's multiple comparison test showed no significant difference between the means of cystatin C, creatinine or urea in the samples taken 60, 90 and 120 minutes after the start of the investigation in the abdominal aorta and the hepatic vein in the healthy volunteers or in the patients suspected of chronic intestinal ischemia with normal angiography. There was no indication of hepatic elimination of cystatin C, creatinine or urea. The serum concentrations of cystatin C, creatinine and urea in the central hepatic vein and the abdominal aorta were independent of the splanchnic blood flow.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
Harnstoff, analytical standard
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
USP
Harnstoff, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
DL-Alanin, ≥99% (HPLC)
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Millipore
Harnstoff -Lösung, suitable for microbiology, 40% in H2O
Sigma-Aldrich
DL-Alanin, ≥99%, FCC, FG
Alanin, European Pharmacopoeia (EP) Reference Standard
Harnstoff, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Harnstoff-12C, 99.9 atom % 12C