Merck
  • Gene expression analyses of the spatio-temporal relationships of human medulloblastoma subgroups during early human neurogenesis.
Gene expression analyses of the spatio-temporal relationships of human medulloblastoma subgroups during early human neurogenesis.
PloS one (2014-11-21)
Cornelia M Hooper, Susan M Hawes, Ursula R Kees, Nicholas G Gottardo, Peter B Dallas
ZUSAMMENFASSUNG
Medulloblastoma is the most common form of malignant paediatric brain tumour and is the leading cause of childhood cancer related mortality. The four molecular subgroups of medulloblastoma that have been identified - WNT, SHH, Group 3 and Group 4 - have molecular and topographical characteristics suggestive of different cells of origin. Definitive identification of the cell(s) of origin of the medulloblastoma subgroups, particularly the poorer prognosis Group 3 and Group 4 medulloblastoma, is critical to understand the pathogenesis of the disease, and ultimately for the development of more effective treatment options. To address this issue, the gene expression profiles of normal human neural tissues and cell types representing a broad neuro-developmental continuum, were compared to those of two independent cohorts of primary human medulloblastoma specimens. Clustering, co-expression network, and gene expression analyses revealed that WNT and SHH medulloblastoma may be derived from distinct neural stem cell populations during early embryonic development, while the transcriptional profiles of Group 3 and Group 4 medulloblastoma resemble cerebellar granule neuron precursors at weeks 10-15 and 20-30 of embryogenesis, respectively. Our data indicate that Group 3 medulloblastoma may arise through abnormal neuronal differentiation, whereas deregulation of synaptic pruning-associated apoptosis may be driving Group 4 tumorigenesis. Overall, these data provide significant new insight into the spatio-temporal relationships and molecular pathogenesis of the human medulloblastoma subgroups, and provide an important framework for the development of more refined model systems, and ultimately improved therapeutic strategies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumacetat, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumacetat, ACS reagent, ≥99.0%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Natriumacetat, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
SAFC
L-Glutamin
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Selen, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Natriumacetat, 99.995% trace metals basis
Sigma-Aldrich
Natriumacetat, >99%, FG
Sigma-Aldrich
Natriumacetat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumacetat, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Natriumacetat -Lösung, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Natriumacetat, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Natriumacetat, meets USP testing specifications, anhydrous
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Selen, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selen, pellets, <5 mm, ≥99.99% trace metals basis
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumacetat, BioXtra, ≥99.0%
USP
Natriumacetat, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Selen, foil, 25x25mm, thickness 3mm, 99.95%
Selen, pellets, < 5mm, ≥99.999%