Merck
  • Netrin-1 promotes medulloblastoma cell invasiveness and angiogenesis, and demonstrates elevated expression in tumor tissue and urine of patients with pediatric medulloblastoma.
Netrin-1 promotes medulloblastoma cell invasiveness and angiogenesis, and demonstrates elevated expression in tumor tissue and urine of patients with pediatric medulloblastoma.
Cancer research (2014-05-09)
Tomoshige Akino, Xuezhe Han, Hironao Nakayama, Brendan McNeish, David Zurakowski, Akiko Mammoto, Michael Klagsbrun, Edward Smith
ZUSAMMENFASSUNG
Invasion and dissemination of medulloblastoma within the central nervous system is the principal factor predicting medulloblastoma treatment failure and death. Netrin-1 is an axon guidance factor implicated in tumor and vascular biology, including in invasive behaviors. We found that exogenous netrin-1 stimulated invasion of human medulloblastoma cells and endothelial cells in contrast to VEGF-A, which promoted invasion of endothelial cells but not medulloblastoma cells. Furthermore, medulloblastoma cells expressed endogenous netrin-1 along with its receptors, neogenin and UNC5B. Blockades in endogenous netrin-1, neogenin, or UNC5B reduced medulloblastoma invasiveness. Neogenin blockade inhibited netrin-1-induced endothelial cells tube formation and recruitment of endothelial cells into Matrigel plugs, two hallmarks of angiogenesis. In patients with pediatric medulloblastoma, netrin-1 mRNA levels were increased 1.7-fold in medulloblastoma tumor specimens compared with control specimens from the same patient. Immunohistochemical analyses showed that netrin-1 was elevated in medulloblastoma tumors versus cerebellum controls. Notably, urinary levels of netrin-1 were 9-fold higher in patients with medulloblastoma compared with control individuals. Moreover, urinary netrin-1 levels were higher in patients with invasive medulloblastoma compared with patients with noninvasive medulloblastoma. Finally, we noted that urinary netrin-1 levels diminished after medulloblastoma resection in patients. Our results suggest netrin-1 is a candidate biomarker capable of detecting an invasive, disseminated phenotype in patients with medulloblastoma and predicting their disease status.