Merck
  • Delivery of glutamine synthetase gene by baculovirus vectors: a proof of concept for the treatment of acute hyperammonemia.
Delivery of glutamine synthetase gene by baculovirus vectors: a proof of concept for the treatment of acute hyperammonemia.
Gene therapy (2014-10-24)
M A Torres-Vega, R Y Vargas-Jerónimo, A G Montiel-Martínez, R M Muñoz-Fuentes, A Zamorano-Carrillo, A R Pastor, L A Palomares
ZUSAMMENFASSUNG
Hyperammonemia, a condition present in patients with urea cycle disorders (UCDs) or liver diseases, can cause neuropsychiatric complications, which in the worst cases result in brain damage, coma or death. Diverse treatments exist for the treatment of hyperammonemia, but they have limited efficacy, adverse effects and elevated cost. Gene therapy is a promising alternative that is explored here. A baculovirus, termed Bac-GS, containing the glutamine synthetase (GS) gene was constructed for the in vitro and in vivo treatment of hyperammonemia. Transduction of MA104 epithelial or L6 myoblast/myotubes cells with Bac-GS resulted in a high expression of the GS gene, an increase in GS concentration, and a reduction of almost half of exogenously added ammonia. When Bac-GS was tested in an acute hyperammonemia rat model by intramuscularly injecting the rear legs, the concentration of ammonia in blood decreased 351 μM, in comparison with controls. A high GS concentration was detected in gastrocnemius muscles from the rats transduced with Bac-GS. These results show that gene delivery for overexpressing GS in muscle tissue is a promising alternative for the treatment of hyperammonemia in patients with acute or chronic liver diseases and hepatic encephalopathy or UCD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ammoniumacetat, ACS reagent, ≥97%
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ammoniak -Lösung, 7 N in methanol
Sigma-Aldrich
Ammoniumacetat, ≥99.99% trace metals basis
Supelco
Ammoniumacetat, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Natriumbutyrat, 98%
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammoniak -Lösung, 2.0 M in ethanol
Sigma-Aldrich
Natriumbutyrat, ≥98.5% (GC)
Sigma-Aldrich
Ammoniak, anhydrous, ≥99.98%
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in dioxane
Sigma-Aldrich
Ammoniak -Lösung, 4 M in methanol
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in THF
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%